Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506160.2 | XP_003506208.1 | 4 - 5 | 595 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Probe spans exons |