Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Steap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506277.2 | XP_003506325.1 | 2 - 3 | 510 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |