Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007642866.1 | XP_007641056.1 | 3 - 4 | 312 | 76 | |
GenBank mRNA | JI883605.1 | - | 1 - 2 | 93 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |