Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007642866.1 | XP_007641056.1 | 4 - 5 | 894 | 56 | |
GenBank mRNA | JI883605.1 | - | 2 - 3 | 675 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |