Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubox5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007637035.1 | XP_007635225.1 | 3 - 4 | 1463 | 63 | |
XM_007637045.1 | XP_007635235.1 | 4 - 5 | 1563 | 63 | ||
XM_007637053.1 | XP_007635243.1 | 4 - 5 | 1574 | 63 | ||
XM_007637062.1 | XP_007635252.1 | 5 - 6 | 1628 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |