Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agtr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508927.1 | XP_003508975.1 | 1 | 522 | 72 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |