Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irak1bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506706.2 | XP_003506754.1 | 2 - 3 | 441 | 140 | |
XM_007647251.1 | XP_007645441.1 | 2 - 3 | 441 | 140 | ||
GenBank mRNA | JP058424.1 | - | 2 - 3 | 158 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |