Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fosl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007650039.1 | XP_007648229.1 | 3 - 4 | 670 | 67 | |
GenBank mRNA | JI867458.1 | - | 2 - 3 | 260 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |