Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500313.2 | XP_003500361.1 | 5 - 6 | 1623 | 95 | |
XM_007641033.1 | XP_007639223.1 | 5 - 6 | 1402 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Transporter 3' Most Probe spans exons |