Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr3g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504392.2 | XP_003504440.1 | 6 - 7 | 497 | 100 | |
XM_007644916.1 | XP_007643106.1 | 5 - 6 | 482 | 100 | ||
GenBank mRNA | JI896141.1 | - | 5 - 6 | 477 | 100 | |
JP055460.1 | - | 5 - 6 | 477 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |