Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507430.1 | XP_003507478.1 | 1 - 2 | 341 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |