Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495750.1 | XP_003495798.1 | 4 - 5 | 459 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |