Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pusl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509966.2 | XP_003510014.2 | 5 - 6 | 723 | 65 | |
GenBank mRNA | JI891969.1 | - | 2 - 3 | 320 | 65 | |
JP054668.1 | - | 2 - 3 | 320 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons |