Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Napsa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510897.1 | XP_003510945.1 | 4 - 5 | 452 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Protease Detect Genomic DNA Probe spans exons |