Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501113.2 | XP_003501161.1 | 9 - 10 | 1172 | 88 | |
GenBank mRNA | JI869123.1 | - | 3 - 4 | 253 | 88 | |
JP063074.1 | - | 3 - 4 | 253 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Transporter Detect Genomic DNA Probe spans exons |