Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pglyrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003505480.2 | XP_003505528.1 | 2 - 3 | 201 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Apoptosis 3' Most Detect Genomic DNA Probe spans exons |