Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scn2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003515274.1 | XP_003515322.1 | 2 - 3 | 238 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Ion Channel Probe spans exons |