Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007630282.1 | XP_007628472.1 | 3 - 4 | 422 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |