Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007644415.1 | XP_007642605.1 | 1 - 2 | 100 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |