Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502640.2 | XP_003502688.1 | 4 - 5 | 347 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |