Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdfi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508996.1 | XP_003509044.1 | 5 - 6 | 790 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Probe spans exons |