Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510537.2 | XP_003510585.1 | 2 - 3 | 324 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |