Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grin3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643247.1 | XP_007641437.1 | 4 - 5 | 2441 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Ion Channel Detect Genomic DNA Probe spans exons |