Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510864.1 | XP_003510912.1 | 4 - 5 | 476 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |