Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496286.2 | XP_003496334.2 | 3 - 4 | 317 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |