Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007650953.1 | XP_007649143.1 | 6 - 7 | 779 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Protease Recommended Assay Probe spans exons |