Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amz2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500314.2 | XP_003500362.1 | 2 - 3 | 428 | 111 | |
XM_007641042.1 | XP_007639232.1 | 2 - 3 | 610 | 111 | ||
XM_007641043.1 | XP_007639233.1 | 3 - 4 | 751 | 111 | ||
XM_007641044.1 | XP_007639234.1 | 2 - 3 | 609 | 111 | ||
GenBank mRNA | JI870883.1 | - | 1 - 2 | 280 | 111 | |
JP043669.1 | - | 1 - 2 | 273 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |