Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdh17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007641223.1 | XP_007639413.1 | 3 - 4 | 2794 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |