Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Heatr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003505142.2 | XP_003505190.1 | 7 - 8 | 2758 | 81 | |
XM_007645667.1 | XP_007643857.1 | 7 - 8 | 2758 | 81 | ||
XM_007645668.1 | XP_007643858.1 | 6 - 7 | 1790 | 81 | ||
XM_007645669.1 | XP_007643859.1 | 7 - 8 | 1848 | 81 | ||
XM_007645670.1 | XP_007643860.1 | 7 - 8 | 1814 | 81 | ||
XM_007645671.1 | XP_007643861.1 | 7 - 8 | 1892 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |