Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509958.2 | XP_003510006.1 | 4 - 5 | 568 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Probe spans exons |