Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502334.1 | XP_003502382.1 | 9 - 10 | 893 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |