Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ikbip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509492.2 | XP_003509540.1 | 1 - 2 | 249 | 105 | |
XM_003509493.2 | XP_003509541.1 | 1 - 2 | 249 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |