Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dexi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496705.2 | XP_003496753.1 | 1 - 2 | 411 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |