Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Narg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513659.2 | XP_003513707.1 | 7 - 8 | 998 | 67 | |
XM_007653918.1 | XP_007652108.1 | 7 - 8 | 928 | 67 | ||
XM_007653919.1 | XP_007652109.1 | 7 - 8 | 1095 | 67 | ||
XM_007653920.1 | XP_007652110.1 | 4 - 5 | 416 | 67 | ||
GenBank mRNA | JI891276.1 | - | 7 - 8 | 955 | 67 | |
JP044394.1 | - | 7 - 8 | 955 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |