Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arpc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502868.2 | XP_003502916.2 | 2 - 3 | 480 | 90 | |
GenBank mRNA | JI879068.1 | - | 2 - 3 | 257 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |