Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003505796.2 | XP_003505844.1 | 6 - 7 | 824 | 78 | |
XM_007646286.1 | XP_007644476.1 | 7 - 8 | 926 | 78 | ||
XM_007646287.1 | XP_007644477.1 | 6 - 7 | 839 | 78 | ||
XM_007646288.1 | XP_007644478.1 | 6 - 7 | 803 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Ion Channel Probe spans exons |