Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyve1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007648884.1 | XP_007647074.1 | 2 - 3 | 258 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Probe spans exons |