Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thtpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502072.2 | XP_003502120.1 | 1 - 2 | 994 | 98 | |
XM_007642685.1 | XP_007640875.1 | 2 - 3 | 636 | 98 | ||
XM_007642686.1 | XP_007640876.1 | 2 - 3 | 778 | 98 | ||
XM_007642687.1 | XP_007640877.1 | 2 - 3 | 895 | 98 | ||
XM_007642688.1 | XP_007640878.1 | 2 - 3 | 575 | 98 | ||
GenBank mRNA | JI864644.1 | - | 1 - 2 | 958 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |