Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnpy4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510420.2 | XP_003510468.1 | 5 - 6 | 684 | 145 | |
XM_007651042.1 | XP_007649232.1 | 5 - 6 | 583 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |