Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lsm8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007645806.1 | XP_007643996.1 | 2 - 3 | 291 | 98 | |
GenBank mRNA | JI863875.1 | - | 2 - 3 | 285 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |