Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sytl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495358.2 | XP_003495406.1 | 13 - 14 | 1740 | 81 | |
XM_003495359.2 | XP_003495407.1 | 12 - 13 | 1674 | 81 |