Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ogfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643787.1 | XP_007641977.1 | 3 - 4 | 520 | 121 | |
GenBank mRNA | JI896721.1 | - | 3 - 4 | 332 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Probe spans exons |