Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bola1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498630.2 | XP_003498678.1 | 2 | 181 | 70 | |
XM_007630820.1 | XP_007629010.1 | 2 | 120 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |