Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INVS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013966062.2 | XP_013821516.1 | 5 - 6 | 828 | 73 | |
XM_013966063.2 | XP_013821517.1 | 5 - 6 | 822 | 73 | ||
XM_018052243.1 | XP_017907732.1 | 4 - 5 | 682 | 73 | ||
XM_018052244.1 | XP_017907733.1 | 4 - 5 | 568 | 73 | ||
GenBank mRNA | JO425249.1 | - | 5 - 6 | 860 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |