Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2W | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005689071.3 | XP_005689128.2 | 4 - 5 | 461 | 125 | |
GenBank mRNA | JO605860.2 | - | 2 - 3 | 1039 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |