Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TACR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005699135.3 | XP_005699192.1 | 1 - 2 | 845 | 99 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Capra hircus Probe spans exons |