Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005684327.3 | XP_005684384.1 | 2 - 3 | 450 | 65 | |
GenBank mRNA | JP351500.2 | - | 2 - 3 | 476 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |