Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDZK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677746.3 | XP_005677803.2 | 7 - 8 | 1260 | 79 | |
XM_005677748.3 | XP_005677805.2 | 6 - 7 | 1138 | 79 | ||
XM_005677749.3 | XP_005677806.2 | 6 - 7 | 1157 | 79 | ||
XM_005677750.3 | XP_005677807.2 | 6 - 7 | 1009 | 79 | ||
XM_013962467.2 | XP_013817921.2 | 6 - 7 | 1087 | 79 | ||
GenBank mRNA | JO428262.1 | - | 7 - 8 | 1505 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |