Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018060223.1 | XP_017915712.1 | 2 - 3 | 287 | 69 | |
GenBank mRNA | JO433359.2 | - | 2 - 3 | 282 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |