Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RILPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005691359.3 | XP_005691416.3 | 5 - 6 | 1229 | 81 | |
XM_013970618.2 | XP_013826072.2 | 5 - 6 | 1233 | 81 | ||
XM_013970619.2 | XP_013826073.2 | 5 - 6 | 1234 | 81 | ||
GenBank mRNA | JO427268.1 | - | 1002 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |